2,5-Dimethoxybenzolsulfonylchlorid

Lieferant: Thermo Scientific

TL00217.1 TL00217.10 TL00217.50
MAYBTL00217.1EA 0 EUR
MAYBTL00217.1 MAYBTL00217.10 MAYBTL00217.50
2,5-Dimethoxybenzolsulfonylchlorid
2,5-Dimethoxybenzolsulfonylchlorid
Formel: C₈H₉ClO₄S
Schmelzpunkt: 119…121 °C
MDL Number: MFCD00024872
CAS-Nummer: 1483-28-9
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR