Phenoxyethanol, Supelco®

Lieferant: Merck
Achtung

Synonyme: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether

M-Clarity™ quality level = MQ100

56753-1ML-F 56753-5ML-F
56753-1ML-FEA 27.2 EUR
56753-1ML-F 56753-5ML-F
Phenoxyethanol, Supelco®
Phenoxyethanol
Formel: C₆H₅OCH₂CH₂OH
Siedepunkt: 245 °C (1013 hPa)
Schmelzpunkt: 13 °C
Dichte: 1.1094 g/cm³ (20 °C)
Flammpunkt: 121 °C (closed cup)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002857
CAS-Nummer: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8718648391680628b2d3cf76f5cf26ffb77988ee&vid=3fd8b3273a938553dccb3f2ccd0836ca193f9c17&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR