Phenetol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethoxybenzene, Ethyl phenyl ether, Phenoxyethane

A15501.0F A15501.AE A15501.AP
A15501.0FEA 310 EUR
A15501.0F A15501.AE A15501.AP
Phenetol ≥98%
Phenetol

Purity: 98+% CAS No.: 103-73-1 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 57°(134°F) Refractive Index: 1.5070 Odour: Aromatic Melting Point: -30° Boiling Point: 169-170° Beilstein Reference: 636270 Merck Reference: 14,7229 UN No.: 3271 MDL No.: MFCD00009090

Formel: C₆H₅OC₂H₅
Siedepunkt: 169…170 °C
Schmelzpunkt: –30 °C
Dichte: 0.967
Flammpunkt: 57 °C (134 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00009090
CAS-Nummer: 103-73-1
EINECS: 203-139-7
UN-Nummer: 3271
ADR: 3,III
Merck Index: 14,7229

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR