4-Amino-3-nitrophenol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Hydroxy-2-nitroaniline
M-Clarity™ quality level = MQ100
249319-100G
249319-25G
SIAL249319-100GEA
74
EUR
SIAL249319-100G
SIAL249319-25G
4-Amino-3-nitrophenol, Sigma-Aldrich®
4-Amino-3-nitrophenol
Formel:
C₆H₆N₂O₃ Schmelzpunkt: 149…152 °C |
MDL Number:
MFCD00066310 CAS-Nummer: 610-81-1 EINECS: 210-236-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...