3-Chlor-4-fluorthiophenol

Lieferant: Thermo Scientific

Synonyme: 1-Chloro-2-fluoro-5-mercaptobenzene, 3-Chloro-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzene-1-thiol

TL00315DA TL00315EA
MAYBTL00315DAEA 0 EUR
MAYBTL00315DA MAYBTL00315EA
3-Chlor-4-fluorthiophenol
3-Chlor-4-fluorthiophenol
Formel: C₆H₄ClFS
MDL Number: MFCD00052938
CAS-Nummer: 60811-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR