3-Chlor-4-fluorthiophenol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Chloro-2-fluoro-5-mercaptobenzene, 3-Chloro-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzene-1-thiol
A10856.03
A10856.06
A10856.14
A10856.03EA
75.5
EUR
A10856.03
A10856.06
A10856.14
3-Chlor-4-fluorthiophenol ≥97%
3-Chlor-4-fluorthiophenol
Purity: 97%
CAS No.: 60811-23-6
Molecular Formula: C6H4ClFS
Formula Weight: 162.61
Boiling Point: 68-70°/2mm
MDL No.: MFCD00052938
Formel:
C₆H₄ClFS Siedepunkt: 68…70 °C (2 mmHg) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00052938 CAS-Nummer: 60811-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...