1-Fluor-4-phenoxybenzol 99%, Stäbe
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B21380.06
B21380.14
B21380.22
B21380.06EA
37.5
EUR
B21380.06
B21380.14
B21380.22
1-Fluor-4-phenoxybenzol 99%, Stäbe
1-Fluor-4-phenoxybenzol
Purity: 99%
CAS No.: 330-84-7
Molecular Formula: C12H9FO
Formula Weight: 188.20
Refractive Index: 1.5570
Formel:
C₁₂H₉FO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00055239 CAS-Nummer: 330-84-7 EINECS: 206-357-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...