4-(Trifluormethoxy)thiophenol 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(Trifluoromethoxy)benzenethiol, 4-(Trifluoromethoxy)benzene-1-thiol

L18674.03 L18674.06
L18674.03EA 59.2 EUR
L18674.03 L18674.06
4-(Trifluormethoxy)thiophenol 95%
4-(Trifluormethoxy)thiophenol

Purity: 95% CAS No.: 169685-29-4 Molecular Formula: C7H5F3OS Formula Weight: 194.18 Refractive Index: 1.4750 Boiling Point: 64-66°/15mm UN No.: 2810 MDL No.: MFCD00052312

Formel: C₇H₅F₃OS
Siedepunkt: 64…66 °C (15 mmHg)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00052312
CAS-Nummer: 169685-29-4
UN-Nummer: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR