3,4-Difluorbenzhydrazid

Lieferant: Thermo Scientific

SPB07437.1 SPB07437.10
MAYBSPB07437.1EA 0 EUR
MAYBSPB07437.1 MAYBSPB07437.10
3,4-Difluorbenzhydrazid
3,4-Difluorbenzhydrazid
Formel: C₇H₆F₂N₂O
MDL Number: MFCD00672992
CAS-Nummer: 229957-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR