3-Fluorphenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid
B21247.03
B21247.06
B21247.14
B21247.03EA
27.4
EUR
B21247.03
B21247.06
B21247.14
3-Fluorphenylboronsäure ≥97%
3-Fluorphenylboronsäure
Purity: 97%
CAS No.: 768-35-4
Molecular Formula: C6H6BFO2
Formula Weight: 139.92
Melting Point: 212-215°
Beilstein Reference: 3030632
Formel:
C₆H₆BFO₂ Schmelzpunkt: 212…215 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00236042 CAS-Nummer: 768-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...