2,6-Difluorphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

B22805.03 B22805.06 B22805.14
B22805.03EA 23.2 EUR
B22805.03 B22805.06 B22805.14
2,6-Difluorphenylboronsäure ≥98%
2,6-Difluorphenylboronsäure

Purity: 98% CAS No.: 162101-25-9 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: ca 140° dec. Beilstein Reference: 8545931

Formel: C₆H₅BF₂O₂
Schmelzpunkt: ∼ 140 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00792436
CAS-Nummer: 162101-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR