Phenylsalicylat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Phenyl 2-hydroxybenzoate, Phenyl Salicylate

B20686.22 B20686.36
B20686.22EA 38.5 EUR
B20686.22 B20686.36
Phenylsalicylat ≥99%
Phenylsalicylat

Purity: 99% CAS No.: 118-55-8 Molecular Formula: C13H10O3 Formula Weight: 214.22 Flash Point: 160°(320°F) Physical Form: Crystalline powder Melting Point: 41-45° Boiling Point: 172-174°/12mm Beilstein Reference: 393969 Merck Reference: 14,7310 MDL No.: MFCD00002213

Formel: C₁₃H₁₀O₃
Siedepunkt: 172…174 °C (12 mmHg)
Schmelzpunkt: 41…45 °C
Dichte: 1.307 g/cm³ (20 °C)
Flammpunkt: 160 °C (320 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002213
CAS-Nummer: 118-55-8
EINECS: 204-259-2
UN-Nummer: 3077
ADR: 9,III
Merck Index: 14,7310

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR