Kaliumhydrogenphthalat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
60359-1KG
60359-250G
SIAL60359-1KGEA
424
EUR
SIAL60359-1KG
SIAL60359-250G
Kaliumhydrogenphthalat, Sigma-Aldrich®
Kaliumhydrogenphthalat
Formel:
HOOCC₆H₄COOK Schmelzpunkt: 295…300 °C Dichte: 1.6362 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013070 CAS-Nummer: 877-24-7 EINECS: 212-889-4 Merck Index: 13,07694 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...