5-Aminosalicylsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
M-Clarity™ quality level = MQ200
A79809-5G
A79809-100G
SIALA79809-5GEA
42.6
EUR
SIALA79809-5G
SIALA79809-100G
5-Aminosalicylsäure, Sigma-Aldrich®
5-Aminosalicylsäure
Formel:
C₇H₇NO₃ Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 280 °C Dichte: 1.568 g/cm³ (20 °C) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00007877 CAS-Nummer: 89-57-6 EINECS: 201-919-1 Merck Index: 12,05964 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...