Diethylphthalat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Phthalic acid diethyl ester

A17529.0B A17529.0E A17529.30
A17529.0BEA 47 EUR
A17529.0B A17529.0E A17529.30
Diethylphthalat ≥99%
Diethylphthalat

Purity: 99% CAS No.: 84-66-2 Molecular Formula: C12H14O4 Formula Weight: 222.24 Flash Point: 160°(320°F) Refractive Index: 1.5020 Melting Point: -64° Boiling Point: 296-298° Beilstein Reference: 1912500 Merck Reference: 14,7371 MDL No.: MFCD00009111

Formel: C₁₂H₁₄O₄
Siedepunkt: 296…298 °C
Schmelzpunkt: –64 °C
Dichte: 1.118
Flammpunkt: 160 °C (320 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00009111
CAS-Nummer: 84-66-2
EINECS: 201-550-6
Merck Index: 14,7371
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR