Methylsalicylat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Wintergreen Oil, Methyl 2-hydroxybenzoate

A10578.0E A10578.30 A10578.36
A10578.0EEA 126 EUR
A10578.0E A10578.30 A10578.36
Methylsalicylat ≥98%
Methylsalicylat

Purity: 98% CAS No.: 119-36-8 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 99°(210°F) Refractive Index: 1.5360 Odour: Characteristic Melting Point: -9° Boiling Point: 224° Beilstein Reference: 971516 Merck Reference: 14,6120 MDL No.: MFCD00002214

Formel: 2-(HO)C₆H₄CO₂CH₃
Molekulargewicht: 152.15 g/mol
Siedepunkt: 224 °C
Schmelzpunkt: –9 °C
Dichte: 1.180
Flammpunkt: 99 °C (210 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002214
CAS-Nummer: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR