4-Chlorbenzamid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A13809.09
A13809.18
A13809.30
A13809.09EA
57.1
EUR
A13809.09
A13809.18
A13809.30
4-Chlorbenzamid ≥98%
4-Chlorbenzamid
Purity: 98+%
CAS No.: 619-56-7
Molecular Formula: C7H6ClNO
Formula Weight: 155.58
Melting Point: 176-180°
Beilstein Reference: 774622
Formel:
C₇H₆ClNO Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 176…180 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007993 CAS-Nummer: 619-56-7 EINECS: 210-601-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...