3-Chlorbenzamid

Lieferant: Thermo Scientific

BTB07622.1 BTB07622.50
MAYBBTB07622.1EA 0 EUR
MAYBBTB07622.1 MAYBBTB07622.50
3-Chlorbenzamid
3-Chlorbenzamid
Formel: C₇H₆ClNO
MDL Number: MFCD00014799
CAS-Nummer: 618-48-4
EINECS: 210-554-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR