2-Chlorphenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
B23324.03
B23324.06
B23324.14
B23324.03EA
54.1
EUR
B23324.03
B23324.06
B23324.14
2-Chlorphenylboronsäure ≥97%
2-Chlorphenylboronsäure
Purity: 97%
CAS No.: 3900-89-8
Molecular Formula: C6H6BClO2
Formula Weight: 156.38
Melting Point: ca 95-100°
Beilstein Reference: 3030414
Formel:
C₆H₆BClO₂ Schmelzpunkt: ∼ 95…100 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00674012 CAS-Nummer: 3900-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...