2,4-Dichlorphenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
L01563.03
L01563.06
L01563.03EA
25.8
EUR
L01563.03
L01563.06
2,4-Dichlorphenylboronsäure ≥98%
2,4-Dichlorphenylboronsäure
Purity: 98+%
CAS No.: 68716-47-2
Molecular Formula: C6H5BCl2O2
Formula Weight: 190.82
Melting Point: 242-245°
Beilstein Reference: 4983878
Formel:
C₆H₅BCl₂O₂ Schmelzpunkt: 242…245 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013930 CAS-Nummer: 68716-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...