1-Brom-4-(difluormethoxy)benzol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(Difluoromethoxy)bromobenzene
B20106.06
B20106.14
B20106.06EA
128
EUR
B20106.06
B20106.14
1-Brom-4-(difluormethoxy)benzol ≥97%
1-Brom-4-(difluormethoxy)benzol
Purity: 97%
CAS No.: 5905-69-1
Molecular Formula: C7H5BrF2O
Formula Weight: 223.02
Refractive Index: 1.5010
Boiling Point: 96-98°/15mm
UN No.: 1993
MDL No.: MFCD00221465
Formel:
C₇H₅BrF₂O Siedepunkt: 96…98 °C (15 mmHg) Dichte: 1.631 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00221465 CAS-Nummer: 5905-69-1 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...