2-Bromphenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2-Bromophenyl)boronic acid, Dihydroxy(2-bromophenyl)borane, 2-Bromobenzeneboronic acid, (2-Bromophenyl)metaboric acid
M-Clarity™ Qualitätsstufe = MQ200
473804-25G
473804-5G
SIAL473804-25GEA
142
EUR
SIAL473804-25G
SIAL473804-5G
2-Bromphenylboronsäure, Sigma-Aldrich®
2-Bromphenylboronsäure
Formel:
C₆H₆BBrO₂ Molekulargewicht: 200.83 g/mol |
MDL Number:
MFCD01114672 CAS-Nummer: 244205-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...