4-Nitrophenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid
M-Clarity™ quality level = MQ100
673854-1G
673854-5G
SIAL673854-1GEA
78.4
EUR
SIAL673854-1G
SIAL673854-5G
4-Nitrophenylboronsäure, Sigma-Aldrich®
4-Nitrophenylboronsäure
Formel:
C₆H₆BNO₄ |
MDL Number:
MFCD00161360 CAS-Nummer: 24067-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...