4-Cyanphenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane
M-Clarity™ quality level = MQ100
521418-10G
521418-1G
SIAL521418-10GEA
357
EUR
SIAL521418-10G
SIAL521418-1G
4-Cyanphenylboronsäure, Sigma-Aldrich®
4-Cyanphenylboronsäure
Formel:
C₇H₆BNO₂ Schmelzpunkt: > 300 °C |
MDL Number:
MFCD01318968 CAS-Nummer: 126747-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...