4-Amino-2,2-difluor-1,3-benzodioxol ≥97%
Lieferant: Thermo Fisher Scientific
B22797.03
B22797.06
B22797.03EA
160
EUR
B22797.03
B22797.06
4-Amino-2,2-difluor-1,3-benzodioxol ≥97%
4-Amino-2,2-difluor-1,3-benzodioxol
Purity: 97+%
CAS No.: 106876-54-4
Molecular Formula: C7H5F2NO2
Formula Weight: 173.12
UN No.: 2810
MDL No.: MFCD00792416
Formel:
C₇H₅F₂NO₂ |
MDL Number:
MFCD00792416 CAS-Nummer: 106876-54-4 UN-Nummer: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...