5-Nitroindolin

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,3-Dihydro-5-nitro-(1H)-indole

RH00433.1 RH00433.10 RH00433.50
MAYBRH00433.1EA 0 EUR
MAYBRH00433.1 MAYBRH00433.10 MAYBRH00433.50
5-Nitroindolin
5-Nitroindolin
Formel: C₈H₈N₂O₂
MDL Number: MFCD00005709
CAS-Nummer: 32692-19-6
EINECS: 251-158-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR