Diethylmaleat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate
L03876.06
L03876.30
L03876.0B
L03876.06EA
26.3
EUR
L03876.06
L03876.30
L03876.0B
Diethylmaleat ≥97%
Diethylmaleat
Purity: 97%
CAS No.: 141-05-9
Molecular Formula: C8H12O4
Formula Weight: 172.18
Flash Point: 93°(199°F)
Refractive Index: 1.4400
Odour: Pleasant
Melting Point: -11°
Boiling Point: 221-224°
Beilstein Reference: 1100825
Merck Reference: 14,3123
MDL No.: MFCD00009191
Formel:
C₈H₁₂O₄ Siedepunkt: 221…224 °C Schmelzpunkt: –11 °C Dichte: 1.064 Flammpunkt: 93 °C (199 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009191 CAS-Nummer: 141-05-9 EINECS: 205-451-9 Merck Index: 14,3123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...