3-(3,4,5-Trimethoxyphenyl)propionsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,4,5-Trimethoxyhydrocinnamic acid, 3-(3,4,5-Trimethoxyphenyl)propanoic acid
189160250
189160050
ACRO189160250EA
139
EUR
ACRO189160250
ACRO189160050
3-(3,4,5-Trimethoxyphenyl)propionsäure 98%
3-(3,4,5-Trimethoxyphenyl)propionsäure
Formel:
C₁₂H₁₆O₅ Siedepunkt: 363 °C (1013 hPa) Schmelzpunkt: 100…105 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002775 CAS-Nummer: 25173-72-2 EINECS: 246-706-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 100 to 105 °C |
HPLC | ≥97.5 % |
Dnase,Rnase,Protease activity | none detected |
Phosphate (PO4) | ≤2 ppm |
Sulfate (SO4) | ≤20 ppm |
Iron (Fe) | ≤2 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...