(4-Hydroxy-3-nitrophenyl)essigsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(4-Hydroxy-3-nitrophenyl)acetic acid

A13858.06 A13858.14
A13858.06EA 144 EUR
A13858.06 A13858.14
(4-Hydroxy-3-nitrophenyl)essigsäure ≥99%
(4-Hydroxy-3-nitrophenyl)essigsäure

Purity: 99% CAS No.: 10463-20-4 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 146-148° Beilstein Reference: 2696044

Formel: (C₆H₃(OH)(NO₂))CH₂COOH
Schmelzpunkt: 146…148 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007122
CAS-Nummer: 10463-20-4
EINECS: 233-948-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR