(3-Bromphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(3-Bromophenyl)acetic acid

M-Clarity™ quality level = MQ100

288861-25G 288861-5G
SIAL288861-25GEA 49.4 EUR
SIAL288861-25G SIAL288861-5G
(3-Bromphenyl)essigsäure, Sigma-Aldrich®
(3-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Siedepunkt: 308 °C (1013 hPa)
Schmelzpunkt: 102…103 °C
MDL Number: MFCD00004330
CAS-Nummer: 1878-67-7
EINECS: 217-522-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR