3,3-Dimethylglutarsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,3-Dimethylpentanedioic acid, 3,3-Dimethyl-1,5-pentanedioic acid
116240250
116241000
116245000
ACRO116240250EA
52.7
EUR
ACRO116240250
ACRO116241000
ACRO116245000
3,3-Dimethylglutarsäure 98%
3,3-Dimethylglutarsäure
Formel:
C₇H₁₂O₄ Siedepunkt: 290 °C (760 mmHg) Schmelzpunkt: 98…102 °C Dichte: 1,4278 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002716 CAS-Nummer: 4839-46-7 EINECS: 225-425-0 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 98 to 102 °C |
Water | ≤1 % (K.F.) |
Titration with NaOH | ≥97.5 % |
Phosphate (PO4) | ≤2 ppm |
Sulfate (SO4) | ≤2 ppm |
Iron (Fe) | ≤0.2 ppm |
Heavy metals (as Pb) | ≤0.5 ppm |
Carbon dioxide (CO2) | ≤0.002 % |
KMnO4-reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...