1-Brom-3,3-dimethylbutanon 97+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone
Purity: 97+%
CAS No.: 5469-26-1
Molecular Formula: C6H11BrO
Formula Weight: 179.06
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.4655
Odour: Pungent
Melting Point: -10°
Boiling Point: 189-192°
Beilstein Reference: 506485
UN No.: 2810
MDL No.: MFCD00000206
Formel:
C₆H₁₁BrO Siedepunkt: 189…192 °C Schmelzpunkt: –10 °C Dichte: 1.333 Flammpunkt: 75 °C (167 °F) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00000206 CAS-Nummer: 5469-26-1 EINECS: 226-794-0 UN-Nummer: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...