1-Octansulfonsäure Natriumsalz ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium octane-1-sulphonate

A14292.14 A14292.06 A14292.22
A14292.14EA 161 EUR
A14292.14 A14292.06 A14292.22
1-Octansulfonsäure Natriumsalz ≥99%
1-Octansulfonsäure Natriumsalz

Purity: 99% CAS No.: 5324-84-5 Molecular Formula: CH3(CH2)7SO3Na Formula Weight: 216.28 Melting Point: 300° Beilstein Reference: 3574241


Reagent for HPLC analysis of peptides and proteins

Formel: C₈H₁₇NaO₃S
Schmelzpunkt: >300 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007544
CAS-Nummer: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR