Dimercaprol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Dimercapto-2,3 propanol-1, Dithioglycerine, 2,3-Dimercaptopropanol, BAL, Dimercaprol, 2,3-Dimercaptopropan-1-ol, 2,3-Dimercaptopropane-1-ol

L03953.06 L03953.03
L03953.06EA 138 EUR
L03953.06 L03953.03
Dimercaprol ≥97%
2,3-Dimercapto-1-propanol

Purity: 97% CAS No.: 59-52-9 Molecular Formula: C3H8OS2 Formula Weight: 124.22 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.244 Refractive Index: 1.5740 Sensitivity: Air Sensitive Appearance: Colourless to yellow Odour: Garlic-like Boiling Point: 120°/15mm Beilstein Reference: 1732058 Merck Reference: 14,3209 UN No.: 2810 MDL No.: MFCD00004864


British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.

Formel: C₃H₈OS₂
Molekulargewicht: 124.22 g/mol
Siedepunkt: 120 °C (15 mmHg)
Schmelzpunkt: 77
Dichte: 1.244
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00004864
CAS-Nummer: 59-52-9
EINECS: 200-433-7
UN-Nummer: 2810
ADR: 6.1 III
Merck Index: 13,03234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745c14e3244d75cab02b536ae9d18ec21829aeba&vid=345ebd39845f4c372f03844bee87b4d8c249ec2b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR