1-Dodecanol ≥98,0% ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol

443816-1KG 443816-5L 443816-25KG
443816-1KGEA 266.3 EUR
443816-1KG HONC443816-5L HONC443816-25KG
1-Dodecanol ≥98,0% ACS, Riedel-de Haën™
1-Dodecanol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Formel: H₃C(CH₂)₁₀CH₂OH
Molekulargewicht: 186.34 g/mol
Siedepunkt: 260…262 °C
Schmelzpunkt: 22…26 °C
Flammpunkt: 121 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00004753
CAS-Nummer: 112-53-8
EINECS: 203-982-0
Merck Index: 13,03439

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay (GC) Min. 98.0 %
Water (Karl Fischer) Max. 0.05 %
APHA Max. 10
Vapor Density 7.4 (vs air)
Explosion Limit 4 %
Autoignition Temperature 500 °F
Vapor Pressure 0.1 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR