2-(Dimethylamino)ethanol ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Dimethylaminoethanol, N,N-Dimethylethanolamine, DMAE

B23616.AK B23616.AU
B23616.AKEA 33.3 EUR
B23616.AK B23616.AU
2-(Dimethylamino)ethanol ≥99%
2-(Dimethylamino)ethanol

Purity: 99+% CAS No.: 108-01-0 Molecular Formula: C4H11NO Formula Weight: 89.14 Flash Point: 40°(104°F) Refractive Index: 1.4295 Odour: Ammonia-like Melting Point: -60° Boiling Point: 134-136° Beilstein Reference: 1209235 Merck Reference: 14,2843 UN No.: 2051 MDL No.: MFCD00002846

Formel: C₄H₁₁NO
Siedepunkt: 134…136 °C
Schmelzpunkt: –60 °C
Dichte: 0.886
Flammpunkt: 40 °C (104 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002846
CAS-Nummer: 108-01-0
EINECS: 203-542-8
UN-Nummer: 2051
ADR: 8,II
Merck Index: 14,2843

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR