Natriumtrifluormethansulfonat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Trifluoromethanesulphonic acid sodium salt
358750050
358750250
ACRO358750050EA
56.3
EUR
ACRO358750050
ACRO358750250
Natriumtrifluormethansulfonat 98%
Natriumtrifluormethansulfonat
Formel:
CF₃NaO₃S Schmelzpunkt: 253…255 °C |
MDL Number:
MFCD00061607 CAS-Nummer: 2926-30-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Assay | ≥97.5 % (ex Na) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...