Ethyltrifluormethansulfonat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
246530-1G
246530-25G
246530-5G
SIAL246530-1GEA
35.4
EUR
SIAL246530-1G
SIAL246530-25G
SIAL246530-5G
Ethyltrifluormethansulfonat, Sigma-Aldrich®
Ethyltrifluormethansulfonat
Formel:
C₃H₅F₃O₃S Siedepunkt: 115 °C (1013 hPa) Dichte: 1.374 g/cm³ (0 °C) |
MDL Number:
MFCD00000410 CAS-Nummer: 425-75-2 EINECS: 207-037-3 UN-Nummer: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...