1,1,1,2,2,3,3-Heptafluor-4-iodbutan 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L04173.06
L04173.03
L04173.06EA
172
EUR
L04173.06
L04173.03
1,1,1,2,2,3,3-Heptafluor-4-iodbutan 96%
1,1,1,2,2,3,3-Heptafluor-4-iodbutan
Purity: 96%
CAS No.: 374-98-1
Molecular Formula: C4H2F7I
Formula Weight: 309.95
Flash Point: None
Refractive Index: 1.3625
Boiling Point: 91-92°
Beilstein Reference: 1761927
MDL No.: MFCD00013719
Formel:
C₄H₂F₇I Siedepunkt: 91…92 °C Dichte: 2,054 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013719 CAS-Nummer: 374-98-1 EINECS: 206-779-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...