di-Ammoniumhydrogencitrat

Lieferant: Roth Carl
Achtung

Synonyme: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

0267.1 0267.2 0267.3
ROTH0267.1EA 0 EUR
ROTH0267.1 ROTH0267.2 ROTH0267.3
di-Ammoniumhydrogencitrat
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
Dichte: 1.483 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013068
CAS-Nummer: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR