D-Methionin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH
M-Clarity™ quality level = MQ200
M9375-25G
M9375-5G
SIALM9375-25GEA
188
EUR
SIALM9375-25G
SIALM9375-5G
D-Methionin, Sigma-Aldrich®
D-Methionin
Formel:
C₅H₁₁NO₂S Siedepunkt: 278 °C (1013 hPa) Schmelzpunkt: 136 °C |
MDL Number:
MFCD00002622 CAS-Nummer: 348-67-4 EINECS: 206-483-6 Merck Index: 13,06004 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...