Glycin (H-Gly-OH) ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Aminoacetic acid, Glycocoll, H-Gly-OH

A13816.0C A13816.0E A13816.36
A13816.0CEA 489 EUR
A13816.0C A13816.0E A13816.36
Glycin (H-Gly-OH) ≥99%
Glycin (H-Gly-OH)
Formel: NH₂CH₂COOH
Siedepunkt: 181 °C (1013 hPa)
Schmelzpunkt: ∼ 245 °C (dec.)
Dichte: 1.595
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White
Form crystalline powder
Assay (Non-aqueous acid-base Titration) 98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR