N-Lauroylsarkosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Lauroyl-Sar-OH, 2-(N-Methyldodecanamido)acetic acid, N-Dodecanoylsarcosine

M-Clarity™ Qualitätsstufe = MQ200

L5000-50G L5000-100G
SIALL5000-50GEA 39.8 EUR
SIALL5000-50G SIALL5000-100G
N-Lauroylsarkosin, Sigma-Aldrich®
N-Lauroylsarkosin
Formel: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH
Molekulargewicht: 271.4 g/mol
MDL Number: MFCD00021749
CAS-Nummer: 97-78-9
EINECS: 202-608-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbc6ba106ced89782062d12fb8ef2edf7f1bad23&vid=723e19724ab9a02efbe0a1d09e95c2385f32c7a4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR