N-Lauroylsarkosin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Lauroyl-Sar-OH, 2-(N-Methyldodecanamido)acetic acid, N-Dodecanoylsarcosine
61739-25G
61739-5G
SIAL61739-25GEA
226
EUR
SIAL61739-25G
SIAL61739-5G
N-Lauroylsarkosin, Sigma-Aldrich®
N-Lauroylsarkosin
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH Molekulargewicht: 271.4 g/mol |
MDL Number:
MFCD00021749 CAS-Nummer: 97-78-9 EINECS: 202-608-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...