Diethyladipat ≥99%, ReagentPlus®, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363
M-Clarity™ quality level = MQ200
245720-100G
245720-500G
SIAL245720-100GEA
58.2
EUR
SIAL245720-100G
SIAL245720-500G
Diethyladipat ≥99%, ReagentPlus®, Sigma-Aldrich®
Diethyladipat
Formel:
C₁₀H₁₈O₄ Siedepunkt: 251 °C (lit.) Schmelzpunkt: –20…–19 °C (lit.) Dichte: 1.009 g/mL (25 °C) (lit.) Flammpunkt: 113 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009215 CAS-Nummer: 141-28-6 EINECS: 205-477-0 Merck Index: 13,00163 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...