4'-Nitroacetanilid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(4-Nitrophenyl)acetamide

A13539.06 A13539.14
A13539.06EA 28.4 EUR
A13539.06 A13539.14
4'-Nitroacetanilid ≥98%
4'-Nitroacetanilid

Purity: 98% CAS No.: 104-04-1 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 215-217° Beilstein Reference: 2211962 Merck Reference: 14,6581 MDL No.: MFCD00007303

Formel: C₈H₈N₂O₃
Siedepunkt: 409 °C (1013 hPa)
Schmelzpunkt: 215…217 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007303
CAS-Nummer: 104-04-1
EINECS: 203-169-0
Merck Index: 14,6581

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR