2'-Nitroacetanilid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(2-Nitrophenyl)acetamide

A16677.06 A16677.14
A16677.06EA 32.4 EUR
A16677.06 A16677.14
2'-Nitroacetanilid ≥98%
2'-Nitroacetanilid

Purity: 98+% CAS No.: 552-32-9 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 90-93° Beilstein Reference: 1959178 Merck Reference: 14,6581 MDL No.: MFCD00016991

Formel: C₈H₈N₂O₃
Molekulargewicht: 180.16 g/mol
Schmelzpunkt: 90…93 °C
Dichte: 1.420
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016991
CAS-Nummer: 552-32-9
EINECS: 209-009-6
Merck Index: 14,6581

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR