3-Oxoglutarsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid
Purity: 97%
CAS No.: 542-05-2
Molecular Formula: C5H6O5
Formula Weight: 146.10
Storage Temperature: 2°C to 8°C
Melting Point: ca 132° dec.
Beilstein Reference: 1447081
Merck Reference: 14,68
MDL No.: MFCD00002711
Formel:
C₅H₆O₅ Siedepunkt: 323 °C (1013 hPa) Schmelzpunkt: ∼ 132 °C (dec.) Dichte: 1.55 g/cm³ (20 °C) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00002711 CAS-Nummer: 542-05-2 EINECS: 208-797-9 Merck Index: 14,68 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...