2-(2-Chlorethoxy)ethanol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ quality level = MQ100
162965-250G
162965-50G
SIAL162965-250GEA
128
EUR
SIAL162965-250G
SIAL162965-50G
2-(2-Chlorethoxy)ethanol, Sigma-Aldrich®
2-(2-Chloroethoxy)ethanol
Formel:
C₄H₉ClO₂ Siedepunkt: 79 °C (1013 hPa) Dichte: 1.175 g/cm³ (20 °C) |
MDL Number:
MFCD00002870 CAS-Nummer: 628-89-7 EINECS: 211-059-9 UN-Nummer: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...