4'-Bromacetanilid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L02924.18
L02924.30
L02924.18EA
51.7
EUR
L02924.18
L02924.30
4'-Bromacetanilid ≥98%
4'-Bromacetanilid
Purity: 98%
CAS No.: 103-88-8
Molecular Formula: C8H8BrNO
Formula Weight: 214.06
Odour: Odourless
Melting Point: 166-171°
Beilstein Reference: 2208091
Merck Reference: 14,1399
MDL No.: MFCD00000092
Formel:
H₃CC(O)NH(C₆H₄Br) Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 166…171 °C Dichte: 1,717 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000092 CAS-Nummer: 103-88-8 EINECS: 203-154-9 Merck Index: 14,1399 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...